siRNA Details
Gene Symbol: Pde1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009978.1 | NP_001009978.1 | 4 | 365 | ||
NM_001009979.1 | NP_001009979.1 | 4 | 365 | |||
NM_001159582.1 | NP_001153054.1 | 6 | 867 | |||
NM_016744.4 | NP_058024.2 | 5 | 662 | |||
GenBank mRNA | AF023529.1 | - | 5 | 582 | ||
AF159298.1 | - | 4 | 365 | |||
AK014810.1 | - | 4 | 364 | |||
AK043647.1 | - | 5 | 663 | |||
AK050004.1 | - | 6 | 728 | |||
AY845863.1 | - | 5 | 414 | |||
AY845864.1 | - | 4 | 365 | |||
BC090628.1 | - | 5 | 630 | 6842675 | ||
U56649.1 | - | 6 | 598 |
