siRNA Details
Gene Symbol: Pdgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008808.3 | NP_032834.1 | 4 | 480 | ||
GenBank mRNA | AK017169.1 | - | 2 | 65 | ||
AY324648.1 | - | 4 | 469 | |||
BC003817.1 | - | 4 | 480 | 3495629 | ||
M29464.1 | - | 4 | 360 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_012801, XM_008768940, XM_006248919, XM_006248918, Z14120, and others.
Gene Ontology Categories:
