siRNA Details
Gene Symbol: Pdha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008811.2 | NP_032837.1 | 1 | 1518 | ||
GenBank mRNA | AK006356.1 | - | 1 | 947 | ||
AK076791.1 | - | 1 | 990 | |||
BC100460.1 | - | 1 | 938 | 30918688 | ||
M76728.1 | - | 1 | 1515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
