siRNA Details
Gene Symbol: Padi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011060.4 | NP_035190.3 | 14 | 1663 | ||
GenBank mRNA | AB013849.1 | - | 14 | 1664 | ||
AK220522.1 | - | 13 | 1905 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
