siRNA Details
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080773.2 | NP_001074242.1 | 11 | 1413 | ||
NM_001286662.1 | NP_001273591.1 | 11 | 1331 | |||
NM_011062.4 | NP_035192.2 | 11 | 1413 | |||
GenBank mRNA | AF079535.1 | - | 11 | 1370 | ||
AF086625.1 | - | 11 | 1379 | |||
AF126294.1 | - | 11 | 1354 | |||
AK147149.1 | - | 11 | 1405 | |||
AK147835.1 | - | 11 | 1388 | |||
AK149162.1 | - | 11 | 1347 | |||
AK149289.1 | - | 11 | 1371 | |||
AK162677.1 | - | 11 | 1373 | |||
AK165564.1 | - | 11 | 1351 | |||
AK188106.1 | - | 3 | 381 | |||
AK193731.1 | - | 3 | 347 | |||
AK205439.1 | - | 2 | 326 | |||
AK219503.1 | - | 3 | 340 | |||
AY062008.1 | - | 11 | 1309 | |||
BC146001.1 | - | 11 | 1350 | 40131304 | ||
BC146003.1 | - | 11 | 1350 | 40131306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
