siRNA Details
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080773.2 | NP_001074242.1 | 13 | 1568 | ||
NM_001286662.1 | NP_001273591.1 | 13 | 1486 | |||
NM_011062.4 | NP_035192.2 | 13 | 1568 | |||
GenBank mRNA | AF079535.1 | - | 13 | 1525 | ||
AF086625.1 | - | 13 | 1534 | |||
AF126294.1 | - | 13 | 1509 | |||
AK147149.1 | - | 13 | 1560 | |||
AK147835.1 | - | 13 | 1543 | |||
AK149162.1 | - | 13 | 1502 | |||
AK162677.1 | - | 13 | 1528 | |||
AK165564.1 | - | 13 | 1506 | |||
AY062008.1 | - | 13 | 1464 | |||
BC146001.1 | - | 13 | 1505 | 40131304 | ||
BC146003.1 | - | 13 | 1505 | 40131306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
