siRNA Details
Gene Symbol: Etv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008815.2 | NP_032841.2 | 11 | 1366 | ||
GenBank mRNA | AK144590.1 | - | 11 | 1319 | ||
AK144683.1 | - | 9 | 1082 | |||
AK166136.1 | - | 11 | 1302 | |||
BC068248.1 | - | 3 | 1983 | |||
DQ832277.1 | - | 10 | 1071 |

More Information
Other Species Targeted

Gene Ontology Categories:
