siRNA Details
Gene Symbol: Per2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011066.3 | NP_035196.2 | 6, 7 | 943 | ||
GenBank mRNA | AF035830.1 | - | 6, 7 | 890 | ||
AF036893.1 | - | 6, 7 | 913 | |||
AK044658.1 | - | 6, 7 | 888 | |||
AK122253.1 | - | 6, 7 | 908 | |||
AK159847.1 | - | 6, 7 | 945 | |||
AK165556.1 | - | 6, 7 | 946 | |||
BC116197.1 | - | 1, 2 | 96 |
Gene Symbol: 9830107B12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC128467.1 | - | 96 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
