siRNA Details
Gene Symbol: Pex11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011068.1 | NP_035198.1 | 3 | 612 | ||
GenBank mRNA | AF093669.1 | - | 3 | 612 | ||
AK076776.1 | - | 2 | 602 | |||
AK078954.1 | - | 2 | 445 | |||
AK149387.1 | - | 3 | 610 | |||
BC023439.1 | - | 3 | 585 | 5251140 | ||
BC028786.1 | - | 1 | 1194 |
