siRNA Details
Gene Symbol: Pex16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145122.2 | NP_660104.2 | 9 | 1163 | ||
GenBank mRNA | AK020743.1 | - | 9 | 873 | ||
AK143073.1 | - | 9 | 1165 | |||
AK162716.1 | - | 9 | 863 | |||
BC010822.1 | - | 9 | 904 | 4218547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
