siRNA Details
Gene Symbol: Pfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019410.3 | NP_062283.1 | 2 | 513 | ||
GenBank mRNA | AJ272203.1 | - | 2 | 244 | ||
AK132651.1 | - | 2 | 321 | |||
AK164145.1 | - | 2 | 215 | |||
BC024363.1 | - | 2 | 293 | 5059568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
