siRNA Details
Gene Symbol: Pgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271705.1 | NP_001258634.1 | 2 | 112 | ||
NM_008827.3 | NP_032853.1 | 2 | 445 | |||
GenBank mRNA | AK042891.1 | - | 2 | 429 | ||
AK088943.1 | - | 2 | 428 | |||
BC016567.1 | - | 2 | 417 | 2649577 | ||
X80171.1 | - | 2 | 213 | |||
X96793.1 | - | 2 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
