siRNA Details
Gene Symbol: Pick1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045558.1 | NP_001039023.1 | 10 | 1016 | ||
NM_008837.3 | NP_032863.2 | 10 | 1075 | |||
GenBank mRNA | AK050224.1 | - | 7 | 1841 | ||
AK090155.1 | - | 1008 | ||||
BC023088.1 | - | 9 | 1611 | |||
BC030363.1 | - | 2 | 1320 | |||
BC030880.1 | - | 1 | 1151 | |||
BC030912.1 | - | 1 | 227 | |||
BC040824.1 | - | 1 | 1151 | |||
BC048788.1 | - | 10 | 961 | 6488048 | ||
BE652516.1 | - | 2 | 211 | |||
CD363874.1 | - | 7 | 505 | |||
Z46720.1 | - | 9 | 764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
