siRNA Details
Gene Symbol: Pigf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008838.1 | NP_032864.1 | 4, 5 | 559 | ||
GenBank mRNA | AK132796.1 | - | 4, 5 | 565 | ||
AK171083.1 | - | 4, 5 | 547 | |||
BC028862.1 | - | 4, 5 | 548 | 3673399 | ||
D50264.1 | - | 4, 5 | 559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
