siRNA Details
Gene Symbol: Pigr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011082.3 | NP_035212.2 | 4 | 1018 | ||
GenBank mRNA | AK128944.1 | - | 4 | 1018 | ||
AK142656.1 | - | 4 | 1020 | |||
BC013556.1 | - | 4 | 951 | 3601095 | ||
U06431.1 | - | 4 | 922 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
