siRNA Details
Gene Symbol: Pira4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011091.1 | NP_035221.1 | 518 | |||
GenBank mRNA | U96685.1 | - | 518 |
Gene Symbol: Gm14548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166672.1 | NP_001160144.1 | 4 | 518 |
Gene Symbol: Pira11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011088.1 | NP_035218.1 | 519 | |||
GenBank mRNA | AF041036.1 | - | 510 | |||
AF055896.1 | - | 510 | ||||
BC103528.2 | - | 519 | 40039374 | |||
BC103529.1 | - | 519 | 40039371 | |||
BC139803.1 | - | 519 | 40039373 | |||
BC150155.1 | - | 522 | 40141323 | |||
BC150712.1 | - | 551 | 9087623 |
Gene Symbol: Pira5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U96686.1 | - | 4 | 507 |
Gene Symbol: Pira7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011094.1 | NP_035224.1 | 4 | 546 | ||
GenBank mRNA | U96688.1 | - | 4 | 546 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
