siRNA Details
Gene Symbol: Pou1f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008849.4 | NP_032875.1 | 6 | 780 | ||
GenBank mRNA | AB622207.1 | - | 5 | 924 | ||
BC061213.1 | - | 897 | 30252457 | |||
D12885.1 | - | 898 | ||||
X57512.1 | - | 6 | 780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
