siRNA Details
Gene Symbol: Pitx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042502.2 | NP_001035967.1 | 3 | 1321 | ||
NM_001042504.2 | NP_001035969.1 | 4 | 812 | |||
NM_001286942.1 | NP_001273871.1 | 5 | 911 | |||
NM_001287048.1 | NP_001273977.1 | 3 | 1321 | |||
NM_011098.4 | NP_035228.2 | 5 | 950 | |||
GenBank mRNA | AB006320.1 | - | 3 | 874 | ||
AB006321.1 | - | 5 | 910 | |||
AF048723.1 | - | 4 | 749 | |||
AF048724.1 | - | 3 | 1282 | |||
AF201091.1 | - | 3 | 648 | |||
AM940438.1 | - | 4 | 616 | |||
AM940439.1 | - | 3 | 574 | |||
BC075660.1 | - | 3 | 986 | 30606653 | ||
U70132.1 | - | 2 | 471 | |||
U80010.1 | - | 5 | 885 | |||
U80011.1 | - | 4 | 765 | |||
U80036.1 | - | 4 | 783 |
