siRNA Details
Gene Symbol: Pkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253883.1 | NP_001240812.1 | 6 | 1094 | ||
NM_011099.3 | NP_035229.2 | 6 | 1094 | |||
GenBank mRNA | AK083076.1 | - | 6 | 887 | ||
AK135397.1 | - | 6 | 887 | |||
AK151724.1 | - | 6 | 960 | |||
AK153483.1 | - | 6 | 960 | |||
AK154695.1 | - | 6 | 887 | |||
AK155110.1 | - | 6 | 887 | |||
AK155655.1 | - | 6 | 890 | |||
AK168943.1 | - | 6 | 887 | |||
AK169872.1 | - | 6 | 866 | |||
AK170892.1 | - | 6 | 890 | |||
AK171023.1 | - | 6 | 887 | |||
AK171033.1 | - | 6 | 890 | |||
AK171106.1 | - | 6 | 891 | |||
AK211250.1 | - | 1 | 50 | |||
BC016619.1 | - | 6 | 854 | 3598842 | ||
BC094663.1 | - | 6 | 833 | 6829532 | ||
D38379.1 | - | 6 | 828 | |||
X97047.1 | - | 5 | 805 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
