siRNA Details
Gene Symbol: Prkcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291434.1 | NP_001278363.1 | 7, 8 | 1084 | ||
NM_011102.4 | NP_035232.1 | 7, 8 | 1084 | |||
GenBank mRNA | AK144419.1 | - | 7, 8 | 1086 | ||
BC111807.1 | - | 7, 8 | 971 | 40039740 | ||
L28035.1 | - | 7, 8 | 1162 | |||
X67129.1 | - | 7, 8 | 893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
