siRNA Details
Gene Symbol: Prkch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008856.3 | NP_032882.2 | 8 | 1097 | ||
GenBank mRNA | AK076352.1 | - | 8 | 1296 | ||
AK145817.1 | - | 8 | 1343 | |||
AK155951.1 | - | 9 | 1440 | |||
AK164044.1 | - | 8 | 1296 | |||
BC031121.1 | - | 8 | 1097 | 5059633 | ||
D90242.1 | - | 8 | 1034 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
