siRNA Details
Gene Symbol: Prkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008858.3 | NP_032884.2 | 15 | 2324 | ||
GenBank mRNA | Z34524.1 | - | 15 | 2199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
