siRNA Details
Gene Symbol: Pkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008861.3 | NP_032887.3 | 10 | 2215 | ||
GenBank mRNA | AF014010.1 | - | 10 | 2118 | ||
AK053502.1 | - | 10 | 2120 | |||
AK054397.1 | - | 1 | 1484 | |||
AK128961.1 | - | 11 | 1989 | |||
AK144421.1 | - | 1 | 2143 | |||
BC053058.1 | - | 10 | 2222 | |||
BC062969.1 | - | 10 | 2222 | |||
FJ609779.1 | - | 9 | 1963 | |||
Y13278.1 | - | 10 | 2131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
