siRNA Details
Gene Symbol: Pkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008861.3 | NP_032887.3 | 15 | 2936 | ||
GenBank mRNA | AF014010.1 | - | 15 | 2839 | ||
AK053502.1 | - | 15 | 2841 | |||
AK128961.1 | - | 16 | 2710 | |||
BC053058.1 | - | 15 | 2943 | |||
BC062969.1 | - | 15 | 2943 | |||
CX566027.1 | - | 6 | 663 | |||
FJ609779.1 | - | 14 | 2684 | |||
Y13278.1 | - | 15 | 2852 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
