siRNA Details
Gene Symbol: Pkig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039390.2 | NP_001034479.1 | 5 | 843 | ||
NM_001039391.2 | NP_001034480.1 | 4 | 757 | |||
NM_001164053.1 | NP_001157525.1 | 4 | 852 | |||
NM_001164055.1 | NP_001157527.1 | 3 | 687 | |||
NM_001277096.1 | NP_001264025.1 | 4 | 754 | |||
NM_011106.2 | NP_035236.1 | 4 | 770 | |||
GenBank mRNA | AK164716.1 | - | 4 | 745 | ||
AK168614.1 | - | 4 | 695 | |||
AK213847.1 | - | 1 | 70 | |||
BC026550.1 | - | 4 | 755 | 5006826 | ||
BY617655.1 | - | 1 | 98 | |||
U97170.1 | - | 4 | 774 |

More Information
Other Species Targeted

Gene Ontology Categories:
