siRNA Details
Gene Symbol: Pkig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039390.2 | NP_001034479.1 | 5 | 1044 | ||
NM_001039391.2 | NP_001034480.1 | 4 | 958 | |||
NM_001164053.1 | NP_001157525.1 | 4 | 1053 | |||
NM_001164055.1 | NP_001157527.1 | 3 | 888 | |||
NM_001277096.1 | NP_001264025.1 | 4 | 955 | |||
NM_011106.2 | NP_035236.1 | 4 | 971 | |||
GenBank mRNA | AK164716.1 | - | 4 | 946 | ||
AK168614.1 | - | 4 | 896 | |||
AK213847.1 | - | 1 | 271 | |||
BC026550.1 | - | 4 | 957 | 5006826 | ||
BY617655.1 | - | 1 | 303 | |||
U97170.1 | - | 4 | 975 |

More Information
Other Species Targeted

Gene Ontology Categories:
