siRNA Details
Gene Symbol: Pklr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013631.2 | NP_038659.2 | 1 | 93 | ||
GenBank mRNA | BC152327.1 | - | 1 | 77 | 40141144 | |
BC152542.1 | - | 1 | 77 | |||
BY107231.1 | - | 1 | 61 | |||
D63764.1 | - | 1 | 93 | |||
S79731.1 | - | 1 | 93 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
