siRNA Details
Gene Symbol: Serpine1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC054091.1 | - | 3, 4 | 650 | 30021925 | |
GQ872459.1 | - | 2, 3 | 508 | |||
M33960.1 | - | 3, 4 | 639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
