siRNA Details
Gene Symbol: Serpine1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008871.2 | NP_032897.2 | 2 | 404 | ||
GenBank mRNA | AK040662.1 | - | 2 | 401 | ||
BC054091.1 | - | 2 | 401 | 30021925 | ||
GQ872458.1 | - | 1 | 259 | |||
GQ872459.1 | - | 1 | 259 | |||
M33960.1 | - | 2 | 390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
