siRNA Details
Gene Symbol: Papola | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011112.3 | NP_035242.1 | 7 | 789 | ||
GenBank mRNA | AK028940.1 | - | 7 | 787 | ||
AK090096.1 | - | 7 | 789 | |||
AK132704.1 | - | 7 | 776 | |||
AK134552.1 | - | 7 | 759 | |||
U52197.1 | - | 7 | 672 | |||
U58134.1 | - | 7 | 733 | |||
U58135.1 | - | 6 | 542 |

More Information
Other Species Targeted

Gene Ontology Categories:
