siRNA Details
Gene Symbol: Papola | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011112.3 | NP_035242.1 | 7 | 793 | ||
GenBank mRNA | AK028940.1 | - | 7 | 791 | ||
AK090096.1 | - | 7 | 793 | |||
AK132704.1 | - | 7 | 780 | |||
AK134552.1 | - | 7 | 763 | |||
U52197.1 | - | 7 | 676 | |||
U58134.1 | - | 7 | 737 | |||
U58135.1 | - | 6 | 546 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
