siRNA Details
Gene Symbol: Plcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013829.2 | NP_038857.1 | 25 | 2294 | ||
GenBank mRNA | AF332071.1 | - | 25 | 2294 | ||
AF332072.1 | - | 25 | 2294 | |||
AK133360.1 | - | 2133 | ||||
AK148001.1 | - | 26 | 2482 | |||
BC089521.1 | - | 26 | 2293 | |||
BC129883.1 | - | 25 | 2289 | 40110646 |
