siRNA Details
Gene Symbol: Plcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081456.1 | NP_001074925.1 | 6 | 920 | ||
NM_148937.2 | NP_683739.2 | 6 | 1071 | |||
GenBank mRNA | AK016945.1 | - | 6 | 1068 | ||
AK029561.1 | - | 6 | 1066 | |||
AK039149.1 | - | 6 | 898 | |||
AK140231.1 | - | 6 | 907 | |||
AY033991.1 | - | 6 | 903 | |||
BC066156.1 | - | 6 | 811 | 30099811 |

More Information
Other Species Targeted

Gene Ontology Categories:
