siRNA Details
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164056.1 | NP_001157528.1 | 5 | 556 | ||
NM_008875.4 | NP_032901.2 | 4 | 501 | |||
GenBank mRNA | BC068144.1 | - | 4 | 358 | 30532470 | |
CF727963.1 | - | 4 | 501 | |||
U87868.1 | - | 5 | 539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
