siRNA Details
Gene Symbol: Pa2g4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011119.3 | NP_035249.1 | 3 | 403 | ||
GenBank mRNA | AK002685.1 | - | 3 | 481 | ||
AK076286.1 | - | 3 | 408 | |||
AK144645.1 | - | 3 | 457 | |||
AK166936.1 | - | 3 | 412 | |||
AK168582.1 | - | 3 | 405 | |||
BC003777.1 | - | 3 | 393 | |||
BC005593.1 | - | 3 | 393 | |||
BC028926.1 | - | 3 | 386 | |||
BC037623.1 | - | 3 | 423 | |||
BC046532.1 | - | 3 | 357 | 5358060 | ||
BC086788.1 | - | 2 | 2521 | |||
U43918.1 | - | 3 | 562 | |||
X84789.1 | - | 3 | 468 |

More Information
Other Species Targeted

Gene Ontology Categories:
