siRNA Details
Gene Symbol: Prl7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011120.2 | NP_035250.1 | 4 | 339 | ||
GenBank mRNA | AK005421.1 | - | 4 | 336 | ||
AK005557.1 | - | 4 | 339 | |||
AK019461.1 | - | 4 | 339 | |||
BC099470.1 | - | 4 | 328 | 30787403 | ||
X02594.1 | - | 4 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
