siRNA Details
Gene Symbol: Plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011121.3 | NP_035251.3 | 8 | 1430 | ||
GenBank mRNA | AK134045.1 | - | 8 | 1371 | ||
AK148050.1 | - | 8 | 1344 | |||
AK148076.1 | - | 8 | 1421 | |||
AK164027.1 | - | 8 | 1400 | |||
BC006880.1 | - | 8 | 1382 | 3596381 | ||
L19558.1 | - | 8 | 1437 | |||
U01063.1 | - | 8 | 1351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
