siRNA Details
Gene Symbol: Plp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290561.1 | NP_001277490.1 | 2 | 334 | ||
NM_001290562.1 | NP_001277491.1 | 2 | 334 | |||
NM_011123.3 | NP_035253.1 | 2 | 334 | |||
GenBank mRNA | AK077564.1 | - | 2 | 290 | ||
AK134554.1 | - | 2 | 325 | |||
AK160872.1 | - | 2 | 329 | |||
AK192772.1 | - | 2 | 217 | |||
BC027010.1 | - | 2 | 244 | 5364736 | ||
M14674.1 | - | 1 | 168 | |||
M15442.1 | - | 2 | 313 | |||
M16472.1 | - | 1 | 168 | |||
M20752.1 | - | 1 | 168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
