siRNA Details
Gene Symbol: Ccl21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011124.4 | NP_035254.1 | 3 | 407 | ||
GenBank mRNA | AF001980.1 | - | 3 | 334 | ||
AF006637.1 | - | 3 | 430 | |||
AK144258.1 | - | 3 | 409 | |||
AK155924.1 | - | 3 | 407 | |||
BC025974.1 | - | 3 | 406 | 4208773 | ||
BC028747.1 | - | 3 | 394 | 1196234 | ||
BC038120.1 | - | 3 | 376 | 3372243 |
Gene Symbol: Ccl21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011335.2 | NP_035465.2 | 370 | |||
GenBank mRNA | W67046.1 | - | 3 | 370 |
Gene Symbol: Ccl21c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023052.2 | NP_075539.1 | 3 | 406 | ||
GenBank mRNA | BC087957.1 | - | 3 | 406 | 6750979 | |
U88322.1 | - | 3 | 379 |
Gene Symbol: Gm10591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193668.1 | NP_001180597.1 | 406 |
Gene Symbol: Gm13304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193666.1 | NP_001180595.1 | 406 |
Gene Symbol: Gm1987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193667.1 | NP_001180596.1 | 3 | 409 |
Gene Symbol: Gm21541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270360.1 | NP_001257289.1 | 3 | 406 | ||
GenBank mRNA | BX522471.1 | - | 3 | 427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
