siRNA Details
Gene Symbol: Pml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008884.5 | NP_032910.3 | 8 | 2059 | ||
NM_178087.4 | NP_835188.2 | 9 | 2197 | |||
GenBank mRNA | AK028044.1 | - | 9 | 2199 | ||
AK032799.1 | - | 7 | 1077 | |||
AK137538.1 | - | 9 | 2140 | |||
AK156279.1 | - | 9 | 2351 | |||
BC020990.1 | - | 9 | 2004 | 4188386 | ||
KJ650238.1 | - | 9 | 2101 | |||
U33626.2 | - | 8 | 1956 |
