siRNA Details
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025570.1 | NP_001020741.1 | 2 | 1307 | ||
NM_011127.2 | NP_035257.1 | 2 | 1307 | |||
NM_175686.3 | NP_783617.2 | 2 | 1307 | |||
GenBank mRNA | AB221686.1 | - | 2 | 292 | ||
AK008428.2 | - | 3 | 137 | |||
AK019503.1 | - | 2 | 118 | |||
AK031075.1 | - | 2 | 273 | |||
AK032773.1 | - | 2 | 874 | |||
AK084387.1 | - | 2 | 1325 | |||
AK089006.1 | - | 2 | 1139 | |||
AK165914.1 | - | 2 | 80 | |||
BC092372.1 | - | 2 | 656 | 30533016 | ||
L06502.1 | - | 2 | 472 | |||
U03873.1 | - | 2 | 1313 | |||
X59725.1 | - | 2 | 433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
