siRNA Details
Gene Symbol: Ppp1r14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008889.2 | NP_032915.2 | 2 | 488 | ||
GenBank mRNA | AK141152.1 | - | 2 | 569 | ||
AK165792.1 | - | 2 | 102 | |||
BC082545.1 | - | 2 | 488 | 6820968 | ||
BC125650.1 | - | 2 | 371 | 40130165 | ||
BC125654.1 | - | 2 | 385 | 40130169 | ||
CF739461.1 | - | 2 | 92 | |||
X97490.1 | - | 2 | 551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
