siRNA Details
Gene Symbol: Pnliprp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011128.2 | NP_035258.2 | 11 | 1378 | ||
GenBank mRNA | AK008021.1 | - | 11 | 1381 | ||
AK131882.1 | - | 11 | 1364 | |||
BC094923.1 | - | 11 | 1337 | 6431510 | ||
M30687.1 | - | 11 | 1371 |

More Information
Other Species Targeted

Gene Ontology Categories:
