siRNA Details
Gene Symbol: Pnliprp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011128.2 | NP_035258.2 | 4 | 472 | ||
GenBank mRNA | AK008021.1 | - | 4 | 477 | ||
AK131882.1 | - | 4 | 458 | |||
BC094923.1 | - | 4 | 431 | 6431510 | ||
M30687.1 | - | 4 | 465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
