siRNA Details
Gene Symbol: Pomc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278581.1 | NP_001265510.1 | 4 | 1109 | ||
NM_001278582.1 | NP_001265511.1 | 4 | 1044 | |||
NM_001278583.1 | NP_001265512.1 | 4 | 1014 | |||
NM_001278584.1 | NP_001265513.1 | 3 | 903 | |||
NM_008895.4 | NP_032921.1 | 3 | 933 | |||
GenBank mRNA | AK017492.1 | - | 3 | 929 | ||
AK017581.1 | - | 3 | 898 | |||
AK030714.1 | - | 4 | 1008 | |||
AK077426.1 | - | 3 | 897 | |||
AK133592.1 | - | 3 | 928 | |||
AK133646.1 | - | 3 | 928 | |||
AK133740.1 | - | 3 | 927 | |||
AK133776.1 | - | 3 | 929 | |||
AK133800.1 | - | 3 | 929 | |||
BB022229.2 | - | 1 | 589 | |||
BC061215.1 | - | 3 | 927 | 30253829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
