siRNA Details
Gene Symbol: Pou2af1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011136.2 | NP_035266.1 | 4 | 524 | ||
GenBank mRNA | AK088587.1 | - | 4 | 524 | ||
AK135080.1 | - | 4 | 525 | |||
BC058611.1 | - | 4 | 505 | 30077096 | ||
U43788.1 | - | 4 | 524 | |||
Z54283.1 | - | 4 | 527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
