siRNA Details
Gene Symbol: Pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010127.3 | NP_034257.1 | 5, 6 | 1279 | ||
GenBank mRNA | AB221558.1 | - | 4, 5 | 569 | ||
AK006732.1 | - | 1, 2 | 359 | |||
BC053046.1 | - | 7, 8 | 1109 | |||
BC059830.1 | - | 5, 6 | 1279 | 6410550 | ||
BC085139.1 | - | 10, 11 | 1657 | 6406821 | ||
D13801.1 | - | 5, 6 | 768 | |||
L13763.1 | - | 4, 5 | 602 |
