siRNA Details
Gene Symbol: Pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010127.3 | NP_034257.1 | 3 | 777 | ||
GenBank mRNA | AB221558.1 | - | 2 | 67 | ||
AY259046.1 | - | 7 | 1911 | |||
AY259047.1 | - | 6 | 1657 | |||
BC053046.1 | - | 5 | 607 | |||
BC059830.1 | - | 3 | 777 | 6410550 | ||
BC085139.1 | - | 8 | 1155 | 6406821 | ||
D13801.1 | - | 3 | 266 | |||
L13763.1 | - | 2 | 100 |
