siRNA Details
Gene Symbol: Endou | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168693.1 | NP_001162164.1 | 5 | 626 | ||
NM_008902.3 | NP_032928.1 | 4 | 625 | |||
GenBank mRNA | AK132616.1 | - | 5 | 567 | ||
AK169457.1 | - | 3 | 716 | |||
M95545.1 | - | 4 | 625 |
