siRNA Details
Gene Symbol: Pparg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011146.3 | NP_035276.2 | 4 | 550 | ||
GenBank mRNA | AB644275.1 | - | 5 | 556 | ||
AY208184.1 | - | 4 | 550 | |||
AY243585.1 | - | 4 | 550 | |||
BC021798.1 | - | 4 | 550 | 4191539 | ||
CN701591.1 | - | 5 | 560 | |||
CT010340.1 | - | 4 | 505 | |||
EF062476.1 | - | 4 | 541 | |||
GQ868647.1 | - | 3 | 308 | |||
GQ868648.1 | - | 3 | 288 | |||
GQ868649.1 | - | 3 | 308 | |||
GQ868650.1 | - | 3 | 288 | |||
U01664.1 | - | 5 | 582 | |||
U01841.1 | - | 5 | 905 | |||
U09138.1 | - | 4 | 544 | |||
U10374.1 | - | 3 | 415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
